missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DGK-zeta Antibody, R&D Systems™

Description
DGK-zeta Polyclonal specifically detects DGK-zeta in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | DGK-zeta |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q13574 |
| Gene Alias | DAGK5, DAGK6DAG kinase zeta, DGKZ, DGKzeta, DGK-ZETA, diacylglycerol kinase zeta, diacylglycerol kinase, zeta, diacylglycerol kinase, zeta 104kDa, Diglyceride kinase zeta, EC 2.7.1.107, hDGKzeta |
| Gene Symbols | DGKZ |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?