missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dishevelled-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3316
This item is not returnable.
View return policy
Description
Dishevelled-1 Polyclonal specifically detects Dishevelled-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Dishevelled-1 | |
| Polyclonal | |
| Unconjugated | |
| O14640 | |
| DVL1 | |
| E. coli-derived recombinant human Dishevelled-1 Arg150-Leu215 Accession # O14640 | |
| 100 μg | |
| Primary | |
| Detects human Dishevelled-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) DVL-2 and rhDVL-3 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| dishevelled 1 (homologous to Drosophila dsh), dishevelled, dsh homolog 1 (Drosophila), Dishevelled1, Dishevelled-1, Dsh, DSH homolog 1, DVL, DVL1, DVL1L1, MGC54245, segment polarity protein dishevelled homolog DVL-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1855 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction