missing translation for 'onlineSavingsMsg'
Learn More

Human DLK2/EGFL9 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7116610
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18771408 25 μg 25µg
18750739 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18771408 Supplier R&D Systems Supplier No. MAB97561SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DLK2/EGFL9 Monoclonal specifically detects DLK2/EGFL9 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLK2/EGFL9
Applications ELISA
Classification Monoclonal
Clone 908866R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q6UY11
Gene Alias delta-like 2 homolog (Drosophila), DLK-2, EGFL9, EGF-like protein 9, EGF-like-domain, multiple 9, Epidermal growth factor-like protein 9, MGC111055, MGC2487, protein delta homolog 2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK-293-derived transfected with human DLK2/EGFL9 Asp27-Ser306 Accession # Q6UY11
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 65989
Test Specificity Detects human DLK2/EGFL9 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.