missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DUX4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9535-SP
This item is not returnable.
View return policy
Description
DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| DUX4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 100288687 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 2142A | |
| Western Blot 0.1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10, DUX4L1 | |
| Human DUX4 synthetic peptide | |
| 25 μg | |
| Primary | |
| Detects human DUX4 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction