missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human E6AP/UBE3A Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: Novus Biologicals™ MAB11498-100
This item is not returnable.
View return policy
Description
E6AP/UBE3A Monoclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Western Blot,Simple Western
Specifications
| E6AP/UBE3A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ANCR, ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, EPVE6AP, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, UBE3A, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human E6AP/UBE3A, Met1-Leu875, Accession # Q05086 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Simple Western | |
| 1077332 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
| Q05086 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7337 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction