Learn More
Novus Biologicals™ Human E6AP/UBE3A Antibody, Novus Biologicals™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ MAB11498-SP
 View more versions of this product
Product Code. 30019018
  • 247.00 € / 25µg
Description
E6AP/UBE3A Monoclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Western Blot,Simple Western
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E6AP/UBE3A
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
ANCR, ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, EPVE6AP, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, UBE3A, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human E6AP/UBE3A, Met1-Leu875, Accession # Q05086
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG1
Western Blot, Simple Western
1077332
Western Blot 2 μg/mL, Simple Western 20 μg/mL
Q05086
Mouse
Protein A or G purified from cell culture supernatant
RUO
7337
Human
Purified