missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ECM-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB39371-100
This item is not returnable.
View return policy
Description
ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ECM-1/Secretory Component P85 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ECM1, extracellular matrix protein 1, p85, Secretory Component Glycoprotein, Secretory Component P85 | |
| Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # Q16610 | |
| 100 μg | |
| Primary | |
| Detects human ECM-1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 966231 | |
| Western Blot 2 ug/mL, ELISA | |
| Q16610 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1893 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction