missing translation for 'onlineSavingsMsg'
Learn More

Human EDA/Ectodysplasin Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18777958
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18777958 25 μg 25µg
18190351 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 18777958 Lieferant R&D Systems Lieferanten-Nr. MAB9222SP

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen EDA/Ectodysplasin
Applications ELISA
Classification Monoclonal
Clone 174806
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001005609
Gene Alias ectodysplasin A, ED1, EDAA1, EDAA2, Tabby, XHED, XLHED
Gene Symbols EDA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1896
Test Specificity Detects human EDA/Ectodysplasin (EDA-A1 and EDA-A2) in ELISAs. In sandwich immunoassays, approximately 50% cross-reactivity with recombinant mouse EDA is observed, and no cross-reactivity with recombinant human (rh) TNF-alpha , rhXEDAR, or rhEDAR is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.