missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EMILIN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10590-SP
This item is not returnable.
View return policy
Description
EMILIN2 Monoclonal antibody specifically detects EMILIN2 in Human samples. It is validated for Immunocytochemistry
Specifications
| EMILIN2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 84034 | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 1031401 | |
| Immunocytochemistry 8-25 ug/mL | |
| Elastin microfibril interface-located protein 2, elastin microfibril interfacer 2EMILIN-2, EMIL2, extracellular glycoprotein EMILIN-2, FLJ33200, FOAP-10, Protein FOAP-10 | |
| Human EMILIN2 synthetic peptide | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction