missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endorepellin/Perlecan Antibody, R&D Systems™

Mouse Monoclonal Antibody
211.00 € - 405.00 €
Specifications
| Antigen | Endorepellin/Perlecan/Heparan Sulfate Proteoglycan |
|---|---|
| Clone | 268908 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18763148
![]() |
R&D Systems
MAB2364-SP |
25 μg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18139590
|
R&D Systems
MAB2364 |
100 μg |
405.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Monoclonal specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human samples. It is validated for Western Blot.Specifications
| Endorepellin/Perlecan/Heparan Sulfate Proteoglycan | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P98160 | |
| 3339 | |
| Mouse myeloma cell line NS0-derived recombinant human Endorepellin/Perlecan Arg3684-Ser4391 (Ser4003Thr) Accession # P98160 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Endorepellin/Perlecan in direct ELISAs and Western blots. |
| 268908 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, HSPG2, Perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 | |
| HSPG2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title