missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Endothelial SFM

Product Code. 11540386 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11540386 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11540386 Supplier Gibco™ Supplier No. 11111044
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Endothelial Serum Free Medium (SFM) has been developed to support the long-term propagation of human umbilical vein endothelial cells (HUVEC).

Human Endothelial Serum Free Medium (SFM) has been developed to support the long-term propagation of human umbilical vein endothelial cells (HUVEC). Complete Human Endothelial SFM has been demonstrated to successfully support primary isolation and subsequent secondary growth of HUVEC for ≤15 passages. HUVEC cultured in complete Human Endothelial SFM exhibit the histotypic 'cobblestone' morphology and retain endothelial specific markers including: expression of Factor VIII-related antigen, UEA-1 lectin binding, uptake of DiI-acetylated LDL, vimentin, and IL-1a-induced ICAM-1 expression. Additionally, HUVEC cultured in Human Endothelial SFM have been shown to maintain cAMP and prostacyclin signal transduction systems. Human Endothelial SFM also supports the growth and retention of physiological markers for human umbilical arterial and dermal microvascular endothelial cells.

Human Endothelial SFM is a basal serum-free growth medium and requires supplementation with bFGF (20 ng/mL), EGF (10 ng/mL), and human plasma fibronectin (10 μg/mL).

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HUAEC, HUVEC
Cell Type Endothelial Cells
Classification cGMP, Serum-free, Chemically-defined
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No Antibiotics
Quantity 500 mL
Shipping Condition Room Temperature
Endotoxin Level ≤10 EU/mL, ≤ 10 EU/mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.