missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ENPP-2/Autotaxin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5255
This item is not returnable.
View return policy
Description
ENPP-2/Autotaxin Polyclonal specifically detects ENPP-2/Autotaxin in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ENPP-2/Autotaxin | |
| Polyclonal | |
| Unconjugated | |
| Q13822 | |
| ENPP2 | |
| Mouse myeloma cell line NS0-derived recombinant human ENPP-2/Autotaxin Asp49-Ile863 Accession # Q13822 | |
| 100 μg | |
| Primary | |
| Detects human ENPP-2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) ENPP-5 and rhENPP-7 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| ATX, ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, ENPP2, Extracellular lysophospholipase D, Lysophosphatidic Acid, LysoPLD, NPP2, PD-IALPHA, PDNP2, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5168 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction