missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ErbB3/Her3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
318.00 € - 597.00 €
Specifications
| Antigen | ErbB3/Her3 |
|---|---|
| Clone | 66223 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746908
![]() |
R&D Systems
FAB3481A-025 |
25 Tests |
318.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18739176
![]() |
R&D Systems
FAB3481A-100 |
100 Tests |
597.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry.Specifications
| ErbB3/Her3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| ERBB3 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human ErbB3/Her3 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human EGF R is observed. |
| 66223 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| P21860 | |
| 2065 | |
| Mouse myeloma cell line NS0-derived recombinant human ErbB3/Her3 Ser20-Thr643 Accession # P21860 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title