missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2873-SP
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Immunohistochemistry.
Specifications
| Erythropoietin/EPO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein, Ala28-Arg193, Accession # CAA26094 | |
| 25 μg | |
| 2056.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 971007 | |
| Immunohistochemistry 5-25 μg;/mL | |
| CAA26094 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Erythropoietin/EPO in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction