missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Exostosin-like 3/EXTL3 Lumenal Domain Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2635
This item is not returnable.
View return policy
Description
Exostosin-like 3/EXTL3 Polyclonal specifically detects Exostosin-like 3/EXTL3 in Human samples. It is validated for Western Blot.
Specifications
| Exostosin-like 3/EXTL3 | |
| Polyclonal | |
| Unconjugated | |
| O43909 | |
| EXTL3 | |
| Mouse myeloma cell line NS0-derived recombinant human Exostosin-like 3/EXTL3 Lumenal Domain Thr52-Ile919 Accession # O43909 | |
| 100 μg | |
| Primary | |
| Detects human Exostosin-like 3/EXTL3 Lumenal Domain in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| botv, DKFZp686C2342, EC 2.4.1.223, exostoses (multiple)-like 3, Exostosin like 3, exostosin-like 3, EXTL1L, EXTL3, EXTR1BOTV, EXT-related protein 1, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Hereditary multiple exostoses gene isolog, KIAA0519, Multiple exostosis-like protein 3, Putative tumor suppressor protein EXTL3, REG, reg receptor, REGR, RPR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2137 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction