missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FABP1/L-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB29642-SP
This item is not returnable.
View return policy
Description
FABP1/L-FABP Monoclonal specifically detects FABP1/L-FABP in Human samples. It is validated for ELISA.
Specifications
| FABP1/L-FABP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
| E. coli-derived recombinant human FABP1/L-FABP Met1-Ile127 Accession # P07148 | |
| 25 μg | |
| 2168 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 965607 | |
| ELISA | |
| P07148 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human FABP1/L-FABP in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction