missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FADD Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2938
This item is not returnable.
View return policy
Description
FADD Polyclonal specifically detects FADD in Human samples. It is validated for Western Blot.
Specifications
| FADD | |
| Polyclonal | |
| Unconjugated | |
| Q13158 | |
| FADD | |
| E. coli-derived recombinant human FADD Met1-Ser208 Accession # Q13158 | |
| 100 μg | |
| Primary | |
| Detects endogenous human FADD in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1, MORT1Fas-associating protein with death domain, protein FADD | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8772 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction