missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc epsilon RI alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
189.00 € - 464.00 €
Specifications
| Antigen | Fc epsilon RI alpha |
|---|---|
| Clone | 773735 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750947
![]() |
R&D Systems
MAB66781-SP |
25 μg |
189.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744597
![]() |
R&D Systems
MAB66781-100 |
100 μg |
464.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc epsilon RI alpha Monoclonal antibody specifically detects Fc epsilon RI alpha in Human samples. It is validated for nanSpecifications
| Fc epsilon RI alpha | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 773735 | |
| Unconjugated | |
| Mouse | |
| Human | |
| Mouse myeloma cell line NS0-derived recombinant human Fc epsilon RI alpha , Val26-Gln205, Accession # P12319 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title