missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RI/CD64 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB12572-SP
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Knockout Validated.
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Fc gamma RI/CD64 in direct ELISAs. | |
| Human | |
| Supernatant |
| Flow Cytometry, CyTOF, Immunoassay | |
| 22 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| IFN-y treated monocytes | |
| 25 μg | |
| 2209 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction