Learn More
Human Fc gamma RIII (CD16) Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 6 publications
Brand:  R&D Systems MAB2546-500
 View more versions of this product
Product Code. 18742347
  • 1675.00 € / 500µg
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fc gamma RIII (CD16)
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2214
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Flow Cytometry, CyTOF
245536
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
O75015
FCGR3B
Raised against Fc gamma RIII (CD16).
500 μg
Primary
Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Fc gamma RIIA and recombinant mouse Fc gamma RIII is observed.
Human
Supernatant