missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIII (CD16) Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2546F-025
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry.
Specifications
| Fc gamma RIII (CD16) | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2214 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 245536 | |
| Flow Cytometry 10 uL/10^6 cells | |
| O75015 | |
| FCGR3A | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIA/B (CD16) Thr20-Gln208 Accession # O75015 | |
| 25 Tests | |
| Primary | |
| Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Fc gamma RIIA and recombinant mouse Fc gamma RIII is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction