Learn More
Human Fetuin A/AHSG Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB1184-SP
Product Code. 18748839
  • 209.00 € / 25µg
Description
Fetuin A/AHSG Monoclonal specifically detects Fetuin A/AHSG in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fetuin A/AHSG
Monoclonal
LYOPH
Western Blot 1 ug/mL
P02765
AHSG
Mouse myeloma cell line NS0-derived recombinant human Fetuin A Ala19-Val367 Accession # P02765
25 μg
Primary
Detects recombinant human Fetuin A in direct ELISAs and Western blots. Does not cross-react with 16 other related proteins tested.
Human
Purified
Western Blot
162922
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
A2HS, AHS, AHSG, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
197
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1