Learn More
Human FFAR1/GPR40 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11025-100
 View more versions of this product
Product Code. 18792567
  • 518.00 € / 100µg
Description
FFAR1/GPR40 Monoclonal antibody specifically detects FFAR1/GPR40 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FFAR1/GPR40
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
FFA1, free fatty acid receptor 1, G protein-coupled receptor 40, GPCR40, GPR40, GPR40FFA1R, G-protein coupled receptor 40
Synthetic peptide within aminoacids 1-50 of human FFAR1/GPR40,, Accession # O14842
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2b
Immunohistochemistry
1048036
Immunohistochemistry 3-25 ug/mL
O14842
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2864
Human
Purified