missing translation for 'onlineSavingsMsg'
Learn More

Human FGF acidic/FGF1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

201.00 € - 476.00 €

Specifications

Antigen FGF acidic/FGF1
Clone 939902
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18721087
GREENER_CHOICE
View Documents
R&D Systems
MAB2321-SP
25 μg
201.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18784137
GREENER_CHOICE
View Documents
R&D Systems
MAB2321
100 μg
476.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

FGF acidic/FGF1
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
FGF1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
939902
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human
P05230
2246
E. coli-derived recombinant human FGF acidic/FGF1, Phe16-Asp155, Accession # P05230
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human FGF acidic/FGF1 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.