missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB233-500
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| FGF basic/FGF2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF2 | |
| E. coli-derived recombinant human FGF basic/FGF2 | |
| 500 μg | |
| Primary | |
| Recognizes human and bovine FGF basic/FGF2. In capture ELISAs, shows approximately 0.2% cross-reactivity with bovine FGF acidic, and no cross-reactivity with recombinant human (rh) FGF acidic, rhFGF-4, rhFGF-6, or rhFGF-7. | |
| Human | |
| Purified |
| ELISA | |
| 10060 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2247 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction