missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2333-SP
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunocytochemistry
Specifications
| FGF basic/FGF2/bFGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
| E. coli -derived human FGF basic/FGF2/bFGF, Accession # P09038 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunocytochemistry | |
| 10044 | |
| Immunocytochemistry 5-25 ug/mL | |
| P09038 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2247 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction