missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFBP2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF9349-SP
This item is not returnable.
View return policy
Description
FGFBP2 Polyclonal specifically detects FGFBP2 in Human samples. It is validated for Immunocytochemistry.
Specifications
| FGFBP2 | |
| Polyclonal | |
| Immunocytochemistry 1-15 ug/mL | |
| Q9BYJ0 | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 83888 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunocytochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 37 kDa killer-specific secretory protein, FGF-binding protein 2, FGF-BP2, FGFBP-2, fibroblast growth factor binding protein 2, fibroblast growth factor-binding protein 2, HBp17-RP, HBP17RP, killer-specific secretory protein of 37 kDa, Ksp37, KSP37HBp17-related protein | |
| Chinese hamster ovary cell line CHO-derived recombinant human FGFBP2 Gln20-Gly223 Accession # Q9BYJ0 | |
| 25 μg | |
| Primary | |
| Detects human FGFBP2 in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction