missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibroblast Activation Protein alpha/FAP Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
226.00 € - 2175.00 €
Specifications
| Antigen | Fibroblast Activation Protein alpha/FAP |
|---|---|
| Clone | 427819 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Flow Cytometry, CyTOF, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18703308
![]() |
R&D Systems
MAB3715-SP |
25 μg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790669
![]() |
R&D Systems
MAB3715-100 |
100 μg |
546.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752519
![]() |
R&D Systems
MAB3715-500 |
500 μg |
2175.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| Fibroblast Activation Protein alpha/FAP | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q12884 | |
| 2191 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed. |
| 427819 | |
| Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| FAP | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title