missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibronectin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB1918-SP
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| Fibronectin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, Immunocytochemistry 3-25 ug/mL | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2335 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| P1H11 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FN1 | |
| Human plasma-derived Fibronectin | |
| 25 μg | |
| Primary | |
| Detects human Fibronectin in Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction