missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FOLR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB5646
This item is not returnable.
View return policy
Description
FOLR1 Monoclonal specifically detects FOLR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated, ELISA Capture (Matched Antibody Pair).
Specifications
| FOLR1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P15328 | |
| FOLR1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human FOLR1 Arg25-Met233 Accession # P15328 | |
| 100 μg | |
| Primary | |
| Detects human FOLR1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human FOLR2, 3 or 4 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Immunoassay, ELISA | |
| 548908 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2348 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction