Learn More
Human FoxP3 Buffer Set; BD
Brand:  BD Biosciences 560098
Product Code. 15859928
  • ONLINE EXCLUSIVE PRICE
    196.00 €
    188.75 € / Each
    Save 7.25 €
    3% Off
Description
Human FoxP3 Buffer Set is optimized for use with the FoxP3 mAb clone 259D/C7. It is intented for the fixation and permeabilization of PBMCs (orlysed whole blood) for intracellular staining of Human FoxP3 and surface staining of the appropriate CD markers. See recommended assay procedure for the Human FoxP3 Buffer Set and the FoxP3 conjugates for the proper use and handling of these products.
Kit components:
Human FoxP3 Buffer A (10 X) 100tests
Human FoxP3 Buffer bA (50 X) 100tests
Applications:
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 Tests
Immunology