missing translation for 'onlineSavingsMsg'
Learn More

Human Frk Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18788878
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18788878 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788878 Supplier R&D Systems Supplier No. MAB3766SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Frk Monoclonal specifically detects Frk in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frk
Applications Immunohistochemistry
Classification Monoclonal
Clone 393812
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P42685
Gene Alias EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, Gtk, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5, PTK5 protein tyrosine kinase 5, Rak, RAKPTK5, tyrosine-protein kinase FRK
Gene Symbols FRK
Host Species Rat
Immunogen E. coli-derived recombinant human Frk Met1-Glu107 Accession # P42685
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2444
Test Specificity Detects human Frk in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.