missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FUBI/MNSF beta Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB9036
This item is not returnable.
View return policy
Description
FUBI/MNSF beta/FAU Monoclonal specifically detects FUBI/MNSF beta/FAU in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| FUBI/MNSF beta/FAU | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 40S ribosomal protein S30, asr1, FAU, FAU1, FAU-encoded ubiquitin-like protein, FBR-MuSV-associated ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed(fox derived), FLJ22986, Fub1, Fubi, MNSFbeta, monoclonal nonspecific suppressor factor beta, ribosomal protein S30, RPS30, S30, ubiquitin-like protein fubi and ribosomal protein S30, ubiquitin-like-S30 fusion protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2197 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 861504 | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P35544 | |
| FAU | |
| E.coli-derived recombinant human FUBI/MNSF beta Met1-Gly74 Accession # P35544 | |
| 100 μg | |
| Primary | |
| Detects human FUBI in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction