missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDF-11/BMP-11 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB19584-100
This item is not returnable.
View return policy
Description
GDF-11/BMP-11 Monoclonal specifically detects GDF-11/BMP-11 in Human samples. It is validated for ELISA.
Specifications
| GDF-11/BMP-11 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11 | |
| E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390 | |
| 100 μg | |
| 10220 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 743829R | |
| ELISA | |
| O95390 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human GDF-11/BMP-11 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction