missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDF-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 406.00 €
Specifications
| Antigen | GDF-3 |
|---|---|
| Clone | 878751 |
| Dilution | Neutralization 7.5-15 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18717139
![]() |
R&D Systems
MAB57541-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18731089
![]() |
R&D Systems
MAB57541 |
100 μg |
406.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-3 Monoclonal specifically detects GDF-3 in Human samples. It is validated for Neutralization.Specifications
| GDF-3 | |
| Neutralization 7.5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2 | |
| GDF3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 878751 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NR23 | |
| 9573 | |
| Chinese hamster ovary cell line CHO-derived human recombinant GDF-3/BMP-2 heterodimer Ala251-Gly364 Accession # Q9NR23 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GDF-3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) BMP-2 and rhGDF-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title