missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDNF Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
207.00 € - 597.00 €
Specifications
| Antigen | GDNF |
|---|---|
| Clone | 27165 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745955
![]() |
R&D Systems
MAB2121-100 |
100 μg |
597.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18786055
![]() |
R&D Systems
MAB2121-SP |
25 μg |
207.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDNF Monoclonal antibody specifically detects GDNF in nan samples. It is validated for ImmunocytochemistrySpecifications
| GDNF | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| P39905 | |
| 2668 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 27165 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, HGDNF, HSCR3 | |
| E. coli-derived recombinant human GDNF, Arg109-Ile211, Accession # P39905 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title