missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GITR/TNFRSF18 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB689-100
This item is not returnable.
View return policy
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation-inducible TNFR family receptor, AITR, AITRTNF receptor superfamily activation-inducible protein, CD357, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, TNFRSF18, tumor necrosis factor receptor superfamily, member 18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8784 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, ELISA | |
| 110416 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction 1-2 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9Y5U5 | |
| TNFRSF18 | |
| Mouse myeloma cell line NS0-derived recombinant human GITR/TNFRSF18 Gln26-Glu161 (Thr43Ala) Accession # Q9Y5U5 | |
| 100 μg | |
| Primary | |
| Detects human GITR in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) 4-1BB, recombinant mouse (rm) 4-1BB, rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFAS, rmGITR, rhHVEM, rhLymphotoxin R beta , rmLymphotoxin R beta , rhNGF R, rhOPG, rmOPG, rhRANK, rmRANK, rhTAJ, rhTNF RI, rmTNF RI, rhTNF RII, or rmTNF RII. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction