missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
221.00 € - 527.00 €
Specifications
| Antigen | GLP1 |
|---|---|
| Clone | 970313R |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710447
![]() |
R&D Systems
MAB110299-SP |
25 μg |
221.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783687
![]() |
R&D Systems
MAB110299-100 |
100 μg |
527.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
GLP1 Monoclonal specifically detects GLP1 in Human samples. It is validated for ELISA.Especificaciones
| GLP1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01275 | |
| 2641 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 970313R | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
| Human GLP-1 synthetic peptide, Accession # P01275 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto