missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR6/GRIK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9610-100
This item is not returnable.
View return policy
Description
KLHL24 Monoclonal antibody specifically detects KLHL24 in Human samples. It is validated for Flow Cytometry
Specifications
| KLHL24 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| DRE1, EAA4, GLR6, GluK2, GLUK6, Kainate Receptor Interacting Protein For GluR6, Kainate Receptor-Interacting Protein For GluR6, Kelch-Like 24, Kelch-Like 24 (Drosophila), Kelch-Like Family Member 24, Kelch-Like Protein 24, KLHL24, KRIP6, MRT6, Protein DRE1 | |
| Chinese Hamster Ovary cell line CHO-derived human GluR6/GRIK2 protein, Thr32 - Asn549, Accession # Q13002 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Flow Cytometry | |
| 818235 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Q13002 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54800 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction