missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glutathione S-Transferase mu 1/ GS Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6894
This item is not returnable.
View return policy
Description
Glutathione S-Transferase mu 1/GSTM1 Polyclonal specifically detects Glutathione S-Transferase mu 1/GSTM1 in Human samples. It is validated for Western Blot.
Specifications
| Glutathione S-Transferase mu 1/GSTM1 | |
| Polyclonal | |
| Unconjugated | |
| P09488 | |
| GSTM1 | |
| E. coli-derived recombinant human Glutathione S-Transferase mu 1/GS Met1-Lys218 Accession # P09488 | |
| 100 μg | |
| Primary | |
| Detects human Glutathione S-Transferase mu 1/GS in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, Glutathione STransferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2944 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction