Learn More
Human Glypican 3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB21191-100
 View more versions of this product
Product Code. 18752177
  • 501.00 € / 100µg
Description
Glypican 3 Monoclonal antibody specifically detects Glypican 3 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glypican 3
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GPC3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS
Mouse myeloma cell line NS0-derived recombinant human Glypican 3, Gln25-Val558, Accession # P51654.1
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG1
ELISA
307808
ELISA
P51654.1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2719
Human
Purified