missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gp130 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
214.00 € - 2090.00 €
Specifications
| Antigen | gp130/CD130 |
|---|---|
| Clone | 28126 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.04-0.2 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774299
![]() |
R&D Systems
MAB228-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18733587
![]() |
R&D Systems
MAB228-100 |
100 μg |
517.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18708019
![]() |
R&D Systems
MAB228-500 |
500 μg |
2090.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
| gp130/CD130 | |
| Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.04-0.2 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3572 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 extracellular domain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human gp130 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R, rhIL-4 R, or rhIL-6 R is observed. |
| 28126 | |
| Flow Cytometry, Neutralization, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| IL6ST | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title