missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR107 Antibody, R&D Systems™

Mouse Monoclonal Antibody
176.00 € - 423.00 €
Specifications
| Antigen | GPR107 |
|---|---|
| Clone | 1040717 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753137
![]() |
R&D Systems
MAB10926-SP |
25 μg |
176.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18720557
![]() |
R&D Systems
MAB10926-100 |
100 μg |
423.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR107 Monoclonal antibody specifically detects GPR107 in Human samples. It is validated for Western BlotSpecifications
| GPR107 | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| bA138E2.2, DKFZp667C222, FLJ16312, FLJ20998, FLJ22591, G protein-coupled receptor 107, KIAA1624, KIAA1624GCDRP, Lung seven transmembrane receptor 1, LUSTR1, LUSTR1MGC126118, MGC15440, protein GPR107, RP11-88G17 | |
| Human GPR107 synthetic peptide, Accession # Q5VW38 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1040717 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q5VW38 | |
| 57720 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title