missing translation for 'onlineSavingsMsg'
Learn More

Human GPR158 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7237138
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18704507 25 μg 25µg
18790857 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18704507 Supplier R&D Systems Supplier No. MAB10286SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR158 Monoclonal antibody specifically detects GPR158 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR158
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 1027604
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 25 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q5T848
Gene Alias G protein-coupled receptor 158, KIAA1136, KIAA1136FLJ37801, probable G-protein coupled receptor 158, RP11-59G22.1
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human GPR158 protein, Ala24-Gln411, Accession # Q5T848
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57512
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.