missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR75 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6728-SP
This item is not returnable.
View return policy
Description
GPR75 Monoclonal specifically detects GPR75 in Human samples. It is validated for Immunohistochemistry.
Specifications
| GPR75 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O95800 | |
| GPR75 | |
| NS0 mouse myeloma cell line transfected with human GPR75 Accession # O95800 | |
| 25 μg | |
| Primary | |
| Detects human GPR75 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 689705 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| G protein-coupled receptor 75, GPRchr2, MGC132002, probable G-protein coupled receptor 75, WI31133, WI-31133 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10936 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction