missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GRAIL/RNF128 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6234-SP
This item is not returnable.
View return policy
Description
GRAIL/RNF128 Polyclonal specifically detects GRAIL/RNF128 in Human samples. It is validated for Western Blot.
Specifications
| GRAIL/RNF128 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 6.3.2, EC 6.3.2.-, FLJ23516, Gene related to anergy in lymphocytes protein, ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128, RNF128 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 79589 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8TEB7 | |
| RNF128 | |
| E. coli-derived recombinant human GRAIL/RNF128 Val42-Pro161 Accession # Q8TEB7 | |
| 25 μg | |
| Primary | |
| Detects human GRAIL/RNF128 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction