missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Granulysin Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 439.00 €
Specifications
| Antigen | Granulysin |
|---|---|
| Clone | 786343 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712267
![]() |
R&D Systems
MAB3138-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744067
![]() |
R&D Systems
MAB3138-100 |
100 μg |
439.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISASpecifications
| Granulysin | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| D2S69Elymphocyte-activation gene 2, GNLY, granulysin, LAG2, LAG-2, Lymphokine, Lymphokine LAG-2, NKG5, NKG5519, Protein NKG5, TLA519, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519 | |
| Mouse myeloma cell line NS0-derived recombinant human Granulysin, Arg23-Leu145, Accession # P22749 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 786343 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P22749 | |
| 10578 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title