missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Granzyme K Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
514.00 € - 529.00 €
Specifications
| Antigen | Granzyme K |
|---|---|
| Clone | 2471A |
| Dilution | Intracellular Staining by Flow Cytometry |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18135391
|
R&D Systems
IC10216T-100UG |
100 μg |
529.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18145511
|
R&D Systems
IC10216T-100 |
100 Tests |
514.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granzyme K Monoclonal antibody specifically detects Granzyme K. It is validated for Flow CytometrySpecifications
| Granzyme K | |
| Intracellular Staining by Flow Cytometry | |
| Monoclonal | |
| Purified | |
| RUO | |
| P49863 | |
| 3003 | |
| IgG | |
| Protein A or G purified from cell culture supernatant |
| 2471A | |
| Flow Cytometry | |
| Alexa Fluor 594 | |
| Rabbit | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, granzyme 3, granzyme K, granzyme K (granzyme 3; tryptase II), granzyme K (serine protease, granzyme 3; tryptase II), Granzyme-3, NK-Tryp-2, NK-tryptase-2, PRSS, TRYP2granzyme-3, tryptase II | |
| Synthetic peptide containing human Granzyme K, Accession # P49863 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title