missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HIF-1 alpha/HIF1A Antibody, R&D Systems™

Rabbit Monoclonal Antibody
203.00 € - 471.00 €
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Clone | 2443B |
| Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunocytochemistry 3-25 μg/mL, Knockout Validated |
| Applications | Western Blot, Immunocytochemistry, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18763517
![]() |
R&D Systems
MAB19351-SP |
25 μg |
203.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761757
![]() |
R&D Systems
MAB19351-100 |
100 μg |
471.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
| HIF-1 alpha | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunocytochemistry 3-25 μg/mL, Knockout Validated | |
| Monoclonal | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF 1A, HIF1, HIF1 alpha, HIF-1 alpha, HIF-1a, HIF1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
| E. coli-derived recombinant human HIF-1 alpha , Arg575-Asn826, Accession # Q16665.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human HIF-1 alpha in direct ELISAs. |
| 2443B | |
| Western Blot, Immunocytochemistry, Immunoassay | |
| Unconjugated | |
| Human | |
| Q16665.1 | |
| 3091 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title