missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HVEM/TNFRSF14 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
199.00 € - 459.00 €
Specifications
| Antigen | HVEM/TNFRSF14 |
|---|---|
| Clone | 2742C |
| Dilution | Immunohistochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732787
![]() |
R&D Systems
MAB3562-SP |
25 μg |
199.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741837
![]() |
R&D Systems
MAB3562-100 |
100 μg |
459.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HVEM/TNFRSF14 Monoclonal antibody specifically detects HVEM/TNFRSF14 in Human samples. It is validated for ImmunohistochemistrySpecifications
| HVEM/TNFRSF14 | |
| Immunohistochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
| Human embryonic kidney cell HEK293-derived human HVEM/TNFRSF14, Pro37-Val202, Accession # Q92956 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2742C | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92956 | |
| 8764 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title